Learn More
Thermo Scientific™ Remel™ QC-Slide™ AFB Stain Control
Product Code. p-4526528
Change view
Click to view available options
Quantity:
10 Slides/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12627609 10 Slides/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12627609 Supplier Thermo Scientific™ Supplier No. R40144
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Remel QC-Slide AFB Stain Control is a heat-fixed quality control slide for acid-fast bacilli stain procedures.
Remel QC-Slide AFB Stain Control is a heat-fixed quality control slide for acid-fast bacilli stain procedures.
TRUSTED_SUSTAINABILITY
Specifications
Description AFB Stain Control
Format Glass Slide
Product Type AFB Stain Control
Packaging Type 10 Slide/Pack
Quantity 10 Slides/Pk.