missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Oxford Modified Agar Base

Product Code. 15238977
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 15238977

Brand: Thermo Scientific™ R454232

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selectively isolate Listeria monocytogenes in food samples with Oxford Modified Agar Base.

Selectively isolate Listeria monocytogenes in food samples with Thermo Scientific™ Remel™ Oxford Modified Agar Base (Dehydrated).

In 1989, Curtis described a selective and differential medium to isolate L. monocytogenes from specimens with mixed flora. This is known as the Oxford Agar1. Lee and McClain developed a selective medium to isolate L. monocytogenes from food products using colistin and moxalactam2 in place of the original selective agents2. This medium is recommended by the American Public Health Association (APHA) for the recovery of L. monocytogenes from meat and dairy products3,4.

Listeria monocytogenes is a common contaminant in raw milk, meats, vegetables, seafood and in the food processing environment. The ability to isolate the organism has been impeded in the past by lack of an effective selective medium, as Listeria monocytogenes can be easily overgrown by competing flora.

Use Oxford Modified Agar Base for selective isolation and detection of Listeria monocytogenes from food and environmental samples.

  • Selective: Lithium chloride and colistin inhibit growth of gram-positive and gram-negative organisms.
  • Recommended by the U.S. Department of Agriculture Food Safety Inspection Service and the American Public Health Association (APHA).

Oxford Agar Base Modified contains Columbia Agar Base that provides peptones and serves as the source of essential nutrients. L. monocytogenes hydrolyzes esculin to produce esculetin which combines with ferric ammonium citrate to form a black precipitate in the medium that surrounds the colonies. Lithium chloride and colistin act as selective agents. Moxalactam Selective Supplement (REF R450551) is added to the medium to increase selectivity.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Oxford Agar Base, Modified
Form Powder
Product Type Agar
Quantity 500 g
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.