Learn More
Thermo Scientific™ Dermatophyte Test Medium (DermaTube)
Product Code. 15414924
Change view
Click to view available options
Quantity:
10 x 12 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414924 10 x 12 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15414924 Supplier Thermo Scientific™ Supplier No. R241045
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Facilitate selective and differential isolation of pathogenic fungi from cutaneous sources using Dermatophyte Test Medium (DermaTube).
Facilitate selective and differential isolation of pathogenic fungi from cutaneous sources using Thermo Scientific™ Remel™ Dermatophyte Test Medium (DermaTube). This medium was formulated by Taplin et al. for use in locations where specialized training and microscopic examination is not available1. A pH indicator and three antimicrobial agents are incorporated into the agar to provide a differential and selective medium for isolation of dermatophytes belonging to the genera Microsporum, Trichophyton, or Epidermophyton.
The dermatophytes are fungi that possess keratinolytic properties that enable them to invade skin, nails and hair2. The infections caused by these organisms are commonly referred to as ringworm and are classified by the Latin word tinea followed by the area of the body infected3. Facilitate selective and differential isolation of pathogenic fungi from cutaneous sources using Dermatophyte Test Medium (DermaTube).
  • Ready to Use: Convenience of prepared media.
  • Selective: Cyclohexamide, chloramphenicol, and gentamycin are selective agents which inhibit most saprophytic fungi and many gram-positive and gram-negative bacteria, including some Pseudomonas.
  • Ease to identify: Phenol red is a pH indicator which detects alkaline metabolites produced by dermatophytes, resulting in a red color development of the medium.
This medium is recommended for use in qualitative procedures. This medium contains soy peptones that supply the nitrogen and carbon compounds necessary for the growth of microorganisms. Dextrose is an energy source.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description DermaTube™
Format 27mm x 70mm Tube
Product Type Microbiology Media
Quantity 10 x 12 mL