missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ D/E (Dey-Engley) Neutralizing Broth (Dehydrated)

Product Code. 13295939
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 13295939

Brand: Thermo Scientific™ R453042

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neutralize chemical disinfectants to confirm bacteriostatic and bactericidal activity in environmental sampling procedures with D/E Neutralizing Broth.

Neutralize chemical disinfectants to distinguish bacteriostatic and bactericidal activity in environmental sampling procedures using Thermo Scientific™ Remel™ D/E (Dey-Engley) Neutralizing Broth (Dehydrated). This medium was formulated by Engley and Dey in 1970 for neutralizing and testing antiseptics and disinfectants1. It is recommended in the Compendium of Methods for the Microbiological Examination of Foods by the American Public Health Association (APHA) for environmental sampling2.

Neutralize chemical disinfectants to distinguish bacteriostatic and bactericidal activity in environmental sampling procedures using D/E (Dey-Engley) Neutralizing Broth. The medium can neutralize a range of chemicals including mercurial, iodine or chlorine preparations, quaternary ammonium compounds, formaldehyde and glutaraldehyde.

  • Easy to Identify: Bromocresol Purple is used as an indicator for the fermentation of dextrose by shifting from purple to yellow at acidic pH.
  • Easy to differentiate: Subculture all tubes of D/E Neutralizing Broth to appropriate media such as D/E Agar to observe growth (viable microorganisms).
    • Bacteriostatic substance – Growth from negative D/E Neutralizing Broth tubes.
    • Bactericidal substance – No growth from negative D/E Neutralizing Broth tubes.
  • Recommended in the Compendium of Methods for the Microbiological Examination of Foods by the American Public Health Association (APHA) for environmental sampling2.

This medium contains peptone and yeast extract that provide a source of nitrogen, carbon, and vitamins. Dextrose provides energy and sodium thiosulfate is incorporated to neutralize chlorine and iodine. Polysorbate 80 neutralizes phenols and formalin. Sodium thioglycolate neutralizes mercurials. Sodium bisulfite neutralizes formaldehyde and glutaraldehyde. Lecithin neutralizes quaternary ammonium compounds.. This medium is suitable for environmental sampling where neutralization of the chemical is important to determine its bactericidal activity.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description D/E Neutralizing Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g

Not intended for IVD use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.