Learn More
Thermo Scientific™ Remel™ Calibrated Inoculating Loops
Product Code. p-4520786
Click to view available options
Product Type:
Inoculating Loop
Color:
Yellow
Quantity:
32/Pk.
Unit Size:
Pack of 32
Product Code. 13212713
Brand: Thermo Scientific™ R501511
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ Remel Calibrated Inoculating Loops come in a variety of materials: platinum, twisted nichrome and plastic.Some with color coded with insulated handle.
Thermo Scientific™ Remel Calibrated Inoculating Loops are available in platinum, twisted nichrome and and plastic. Some are color coded with insulated handle.
  • Twisted nichrome or plastic
  • With or without insulated handle
  • Color-coded
TRUSTED_SUSTAINABILITY
Specifications
Volume 10 μL
Material Plastic, Nichrome
Color Yellow
Product Type Inoculating Loop
Quantity 32/Pk.