Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Click to view available options
Description:
Polyvalent, Phase 2
Polyvalent, Phases 1 and 2
Rapid Diagnostic 1 (b, d, E, r)
Rapid Diagnostic 2 (b, E, k, L)
Rapid Diagnostic 3 (d, E, G, k)
Salmonella H (1, 2)
Salmonella H (1, 5)
Salmonella H (1, 6)
Salmonella H (1, 7)
Salmonella H (E) Polyvalent
Salmonella H (G) Polyvalent
Salmonella H (L) Polyvalent
Salmonella H (a)
Salmonella H (b)
Salmonella H (c )
Salmonella H (d)
Salmonella H (eh)
Salmonella H (enx)
Salmonella H (f, g)
Salmonella H (gm)
Salmonella H (i)
Salmonella H (r )
Salmonella H (y)
Salmonella H (z27)
Unit Size:
2mL
Each
Description
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
- Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Specifications
| Control Sets | Positive |
| Description | Salmonella H (E) Polyvalent |
| Detectable Analytes | Salmonella H Antigen |
| Type | Agglutinating Sera |
| Material | Polyvalent |
| Packaging Type | 2 mL/Vial |
| Quantity | 2 mL |
| Target Organism Class | Salmonella H |