Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Product Code. 12617429
Click to view available options
Description:
Polyvalent, Phase 2
Polyvalent, Phases 1 and 2
Rapid Diagnostic 1 (b, d, E, r)
Rapid Diagnostic 2 (b, E, k, L)
Rapid Diagnostic 3 (d, E, G, k)
Salmonella H (1, 2)
Salmonella H (1, 5)
Salmonella H (1, 6)
Salmonella H (1, 7)
Salmonella H (E) Polyvalent
Salmonella H (G) Polyvalent
Salmonella H (L) Polyvalent
Salmonella H (a)
Salmonella H (b)
Salmonella H (c )
Salmonella H (d)
Salmonella H (eh)
Salmonella H (enx)
Salmonella H (f, g)
Salmonella H (gm)
Salmonella H (i)
Salmonella H (r )
Salmonella H (y)
Salmonella H (z27)
Unit Size:
2mL
Each
Product Code. 12617429
Brand: Thermo Scientific™ R30162801
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifications
Control Sets Positive
Description Salmonella H (z27)
Detectable Analytes Salmonella H Antigen
Type Agglutinating Sera
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella H