Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Neisseria meningitidis
Product Code. 13255589
Change view
Click to view available options
Description:
Group A
Group B
Group C
Group D
Group W135
Unit Size:
2mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
13255589 Group C 2mL
13245589 Group A 2mL
11414079 Group D 2mL
13215599 Group B 2mL
13295589 Group W135 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13255589 Supplier Thermo Scientific™ Supplier No. R30166901
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
Used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
Remel Agglutinating Sera are used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
TRUSTED_SUSTAINABILITY
Specifications
Description Group C
CE Marker Yes
Detectable Analytes N. meningitidis
Type Neisseria Meningitidis Test Kit
Organism Group Group C
Quantity Each
Target Organism Class Neisseria Meningitidis