Learn More
Abnova™ RELA/CEN11p FISH Probe
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark |
| Format | Tube |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Gene | v-rel reticuloendotheliosis viral oncogene homolog A (avian) |
| Gene Alias | MGC131774,NFKB3,p65 |
| Gene Symbol | RELA |
| Gene ID (Entrez) | 5970 |
| Product Type | DNA/RNA |
| Quantity | 100 μL |
| Form | Liquid |