Learn More
Fisherbrand™ Reinforced Silicone Tubing
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
3 mm
5 mm
12.5 mm
16.0 mm
Diameter (Metric) Outer:
8.0 mm
11.4 mm
18.9 mm
25.6 mm
Maximum Pressure:
30 bar
40 bar
60 bar
Thickness (Metric):
2.5 mm
3.2 mm
4.8 mm
Unit Size:
Each
Description
- Autoclavable
- Glossy in appearance
- Translucent and flexible
- FDA (food) compliant
- Applications include pharmaceutical, high pressure and temperature work, biotechnology and aerospace
Specifications
Specifications
| Certifications/Compliance | FDA, BFR and WRAS approved |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 16.0 mm |
| Diameter (Metric) Outer | 25.6 mm |
| Overall Length (Metric) | 25 m |
| Thickness (Metric) | 4.8 mm |
| Maximum Pressure | 30 bar |