Learn More
Brady™ Reinforced Polypropylene Warning Sign
Product Code. 10410884
Change view
Click to view available options
Height (Metric):
200 mm
315 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
10410884 315 mm Each
10694725 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10410884 Supplier Brady™ Supplier No. 252201
Only null left
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC 300-TRI 315-B7527
Material Polypropylene
Quantity 1 Ea.
Description Reinforced Polypropylene
For Use With (Application) Variety of applications both indoors and outdoors
Height (Metric) 315 mm
Thickness 0.369 mm
Width (Metric) 315 mm
Temperature Range (Metric) -40°C to 120°C
Shape Triangular
Displayed Text Warning of Flammable Materials
Product Type Warning Sign
Color Black on Yellow