Learn More
Brady™ Reinforced Polypropylene Toxic Substance Warning Signs
Specifications
Specifications
| Alternate Part Number | PIC 302-TRI 100-B7527 |
| Material | Polypropylene |
| Quantity | 1 Ea. |
| Description | Reinforced Polypropylene |
| For Use With (Application) | Safety signs warning of toxic substances |
| Height (Metric) | 100 mm |
| Thickness | 0.369 mm |
| Width (Metric) | 100 mm |
| Temperature Range (Metric) | -40°C to 120°C |
| Shape | Triangular |
| Show More |