Learn More
Brady™ Reinforced Polypropylene Radioactive Warning Sign
Product Code. 10705695
Change view
Click to view available options
Height (Metric):
200 mm
315 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
10705695 200 mm Each
10597234 315 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10705695 Supplier Brady™ Supplier No. 252208
Only null left
Specifications
Alternate Part Number PIC 304-TRI 200-B7527
Material Polypropylene
Quantity 1 Ea.
Description Reinforced Polypropylene
For Use With (Application) Used for indoor and outdoor safety signs
Height (Metric) 200 mm
Max. Temperature (Metric) 120°C
Thickness 0.369 mm
Width (Metric) 200 mm
Shape Triangular
Displayed Text Warning of Radioactive materials or radiation ioniserenden
Product Type Radioactive Warning Sign
Color Black on Yellow