Learn More
Brady™ Reinforced Polypropylene Radiation Laser Sign
Product Code. 11770194
Click to view available options
Unit Size:
Each
Product Code. 11770194
Brand: Brady™ 223480
Only null left
Specifications
Alternate Part Number PIC 304-TRI 100-B7527
Material Polypropylene
Quantity 1 Ea.
Description Reinforced Polypropylene
For Use With (Application) Used for a wide variety of applications both indoors and outdoors
Height (Metric) 100 mm
Max. Temperature (Metric) 120°C
Thickness 0.369 mm
Width (Metric) 100 mm
Shape Triangular
Displayed Text Warning of radioactive materials or radiation ioniserenden
Product Type Radiation Laser Sign
Color Black on Yellow