Learn More
Brady™ Reinforced Polypropylene Mandatory Signs
Brand:  Brady™ 252111
Additional Details : Weight : 0.04300kg
 View more versions of this product
Product Code. 10020624
  • 13.55€ / Each
Estimated Shipment: 13-08-2024
Specifications
Polypropylene
Reinforced Polypropylene
120°C
Round
Mandatory Sign
1 Ea.
200 mm
0.369 mm
use Foot
White on Blue