Learn More
Taylor Wharton™ LS750B Cryostorage System
Click to view available options
Holds:
750 Vials
Unit Size:
Each
Description
- Rugged construction
- Convenient storage system
- Superior vacuum performance
- Lockable lids
- Optional added security
Specifications
Specifications
| Capacity (Metric) | 35 L |
| Closure Type | Lid |
| Type | Cryo Box |
| Holds | 750 Vials |
| Width (Metric) | 478 mm |
| Height (Metric) | 681 mm |