Learn More
Haiminger™ Refrigerator
Product Code. 12644357 Shop All Liebherr Appliances Products
Change view
Click to view available options
Capacity (Metric) Refrigerator:
221 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Refrigerator Unit Size
12644357 221 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12644357 Supplier Haiminger™ Supplier No. K 2330
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Type General Purpose Refrigerator
Capacity (Metric) Refrigerator 221 L
Door Style Reversible
No. of Doors 1
Shelves 5
Defrost Automatic
Casters Yes
Color White
Control Type Mechanical
Model K 2330
Warranty 2 Years
Dimensions (DxWxH) Exterior 1170 H x 550 W x 629 Dmm
Depth (Metric) Exterior 629 mm
Height (Metric) Exterior 1170 mm
Width (Metric) Exterior 555 mm
Energy Usage 0.344kWh/24 hr.
Voltage 220/240 V
Electrical Requirements 220 - 240 V 50 Hz
Frequency 50 Hz
Shipping Weight (Metric) 50 kg
Weight (Metric) 47 kg