Learn More
Abeba™ Reflexor™ 1064 Shoes
Antistatic shoe includes PU-sole with high slip resistance.
Brand:  Abeba™ 1064/37
Additional Details : Weight : 0.70000kg
 View more versions of this product
Product Code. 15686847
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • Smooth leather
  • Black/white
  • Slip resistance PU outsole sole
  • Integrated Reflexor™ massage footbed
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Instep area with elastic band
  • Hand-sewn seam
Specifications
Specifications
Black
Leather
Slip-resistant
1064
Microfiber
37
Polyurethane