Learn More
Zeiss™ Reflector Module Bright Field
Product Code. 11875440
Change view
Click to view available options
Type:
Reflector Module Bright Field ACR P and C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11875440 Reflector Module Bright Field ACR P and C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11875440 Supplier Zeiss™ Supplier No. 4249289901000
This item requires a price update prior to being ordered.
Request a Price for this item here.
Use this reflector module for a variety of reflected light microscopy applications
  • Bright Field Reflector Module
  • Accessory for Zeiss Microscopes
Specifications
Type Reflector Module Bright Field ACR P and C
For Use With (Application) Reflected light microscopy
Quantity 1 Ea.