Learn More
Olympus™ Reflected Light Darkfield Mirror Unit, U-MDF3
Description
- Half-mirror for reflected light darkfield illumination.
- Includes filter mirror set plus blank cube.
- Narrow style for the 6-position turret on the Olympus BX2 and IX2 series microscopes.
- Also retrofits the older 4-position turret on the BX and IX series microscopes.
Specifications
Specifications
| Type | Reflected Light, Darkfield Mirror Unit |
| For Use With (Equipment) | BX2, IX2, BX, IX microscopes |