Learn More
Olympus™ Reflected Light Darkfield Mirror Unit, U-MDF3
Product Code. 12691521
Change view
Click to view available options
For Use With (Equipment):
BX2, IX2, BX, IX microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12691521 BX2, IX2, BX, IX microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12691521 Supplier Olympus™ Supplier No. K10017464
  • ONLINE EXCLUSIVE PRICE
    492.00€
    334.81€ / Pack of 1
    Save 157.19€
    31% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Use this cube-shaped mirror unit for brightfield illumination. Olympus™ Reflected Light Darkfield Mirror Unit, U-MDF3 is a narrow-style for the 6-position turret on the BX and IX series of microscopes.
  • Half-mirror for reflected light darkfield illumination.
  • Includes filter mirror set plus blank cube.
  • Narrow style for the 6-position turret on the Olympus BX2 and IX2 series microscopes.
  • Also retrofits the older 4-position turret on the BX and IX series microscopes.
Specifications
Type Reflected Light, Darkfield Mirror Unit
For Use With (Equipment) BX2, IX2, BX, IX microscopes