Learn More
Labcon™ Refillable Racks for Eclipse™ Refill Tips
GREENER_CHOICE
Product Code. 16631963
Change view
Click to view available options
Holds:
Pipette Tips (10 μL)
Pipette Tips (10 to 1250 μL)
Pipette Tips (10 to 250 μL)
Pipette Tips (100 to 1250 μL)
Pipette Tips (1000 to 1250 μL)
Color:
Assorted
Purple
Teal
Unit Size:
Pack of 10
Pack of 12
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Color unitSize
16631963 Pipette Tips (1000 to 1250 μL) Purple Pack of 6
16641963 Pipette Tips (100 to 1250 μL) Purple Pack of 6
16651963 Pipette Tips (100 to 1250 μL) Assorted Pack of 6
16661963 Pipette Tips (10 to 250 μL) Purple Pack of 10
16671963 Pipette Tips (10 to 1250 μL) Assorted Pack of 10
17567876 Pipette Tips (10 μL) Teal Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16631963 Supplier Labcon™ Supplier No. 12779600069
Estimated Shipment: 01-09-2026
Add to basket
Edge
Add to basket
Each rack is designed to hold one refill of 96 tips.
  • Pipette tip rack
  • Refillable
  • With hinged clear lid
  • For use with Eclipse™ pipette tip refills
  • Holds one refill platform of 96 tips
  • Autoclavable
  • Made in USA
Specifications
Certifications/Compliance ISO 9001:2015, FDA CFR21 GMP
Autoclavable Autoclavable
Color Purple
Holds Pipette Tips (1000 to 1250 μL)
Material Polypropylene
Sterility Non-sterile
For Use With (Application) Liquid Handling
No. per Pack 6/Pk.
Quantity 6 Racks/Pk., 8 Pk./Cs.
Shipping Condition Room Temperature
Product Type Refillable