Description
- QuickTask ReFIBE Mop Heads are manufactured from 100% recycled 2-ply polyester around a recycled polyester core
- These mop heads have been specifically designed for use with Contecs Mop Saturation Systems. With high absorbency the mops are suitable for single use saturation, giving a surface coverage up to approximately 20 sqm.
- Also suitable for the application of disinfectants and removal of light soil, residues and biofilms.
- Designed for single use, the mop heads are compatible with a range of chemicals and disinfectants.
- Made from 100% recycled polyester materials
- 100% recycled 2-ply polyester mop heads
- High sorbency
- Available sterile
- Suitable for hands free mop application with appropriate hardware
- Double bagged
Specifications
Specifications
| Product Type | Mop Pad |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Color-coded | No |
| Material | Polyester |