Learn More
SI Analytics™ Reference Electrode
Product Code. 10024211 Shop All SI Analytics Products
Change view
Click to view available options
Connector Types:
Screw Plug Head
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
10024211 Screw Plug Head Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10024211 Supplier SI Analytics™ Supplier No. 285111767
  • ONLINE EXCLUSIVE PRICE
    278.00€
    262.65€ / Pack of 1
    Save 15.35€
    5% Off
View all promotions
In Stock
Type B 1881 HD: Threaded Screw Plug Connection, PVDF Shaft and Diaphragm, Heavy duty with Ag/AgCl Reference System.
  • Ag / AgCl reference
  • PVDF diaphragm
  • Paste electrolyte
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Screw Plug Head
Material PVDF Shaft
Type Reference Electrode
Temperature (Metric) Range -5°C to +100°C
Length (Metric) 120 mm
Diameter (Metric) Body 12 mm