Learn More
Buerkle™ Reduced Socket with Inner Thread
Shop All Bürkle Products
Click to view available options
Width (Metric):
17.5 mm
23 mm
Unit Size:
Each
Description
- Polypropylene construction
- Autoclavable to 121°C
- Safe for use with foodstuffs
- Supplied in packs of 10
Specifications
Specifications
| Certifications/Compliance | EU Foodstuffs |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Thread Size | 1/2 to 3/4 in. |
| Width (Metric) | 17.5 mm |
| Product Type | Socket Reduced Inner Thread |