Learn More
Brady™ Red and White Warning Tape
Product Code. 11736655
Click to view available options
Dimensions (L x W):
33 m x 38 mm
33 m x 50 mm
33 m x 75 mm
Unit Size:
1 roll
Product Code. 11736655
Brand: Brady™ 255295
Only null left
Add to basket
Add to basket
The marking of transport routes must be both flexible and durable, as they are constantly adapted to the changing demands and needs within your facility or warehouse. Self-adhesive Red and White Warning Tape for marking pillars, pylons, the edges of walls, and gate arches.
  • The marking of transport routes must be both flexible and durable, as they are constantly adapted to the changing demands and needs within your facility or warehouse.
  • Self-adhesive warning film for marking pillars, pylons, the edges of walls, and gate arches.
Specifications
Alternate Part Number 255295
Description Warning Tape - Red / White
Dimensions (L x W) 33 m x 38 mm
For Use With (Application) Marking pillars, pylons, the edges of walls, gate arch, transport routes
Length (English) 108.3 ft.
Length (Metric) 33 m
Packaging Quantity 1 Roll
Thickness 0.138 mm
Width (English) 1.5 in.
Width (Metric) 38 mm
Adhesive Type Self-adhesive
Color Red, White
Type Floor Marking Tape