Learn More
Kautex™ Red Polypropylene Screw Closures
Click to view available options
For Use With (Equipment):
10 to 1000 mL Bottle
100 mL Bottle
1500/2500/4000 mL Bottle
250/350 mL Bottle
50 mL Bottle
500/750/1000 mL Bottle
Diameter (Metric):
28 mm
32 mm
45 mm
54 mm
80 mm
Unit Size:
Each
Pack of 100
Pack of 210
Description
- PP
- Red
- Tamper evident ring
- PE-foam insert
Specifications
Specifications
| Color | Red |
| Includes | Pull-off Ring and Polyethylene Foam Insert |
| Material | Polypropylene |
| Liner Material | Polyethylene Foam Insert |
| Product Type | Screw Cap |
| For Use With (Equipment) | 10 to 1000 mL Bottle |
| Diameter (Metric) | 32 mm |