missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Red-Gal
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Description
An alternative chromogenic substrate for β-D-galactosidase.
Red-GAL is used in conjuction with IPTG for detection of β-galactosidase activity in bacterial colonies in a colorimetric assay to detect recombinants from non-recombinants.
- Substance Name: 6-Chloro-3-indolyl-β-D-galactopyranoside
- CAS Number: 138182-21-5
- Molecular Formula: C14H16CINO6
- Molecular Weight: 329.7
Specifications
Specifications
| Quantity | 100 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction