missing translation for 'onlineSavingsMsg'
Learn More

Melford Red-Gal

Product Code. p-200087279
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 30018155

Brand: Melford C400401.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An alternative chromogenic substrate for β-D-galactosidase.

Red-GAL is used in conjuction with IPTG for detection of β-galactosidase activity in bacterial colonies in a colorimetric assay to detect recombinants from non-recombinants.
  • Substance Name: 6-Chloro-3-indolyl-β-D-galactopyranoside
  • CAS Number: 138182-21-5
  • Molecular Formula: C14H16CINO6
  • Molecular Weight: 329.7

Specifications

Quantity 1 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.