Learn More
Haleco™ Recovery Bags
Product Code. p-8010637
Click to view available options
Color:
Black
Clear
Unit Size:
Pack of 200
Pack of 500
Product Code. 12516239
Brand: Haleco™ SACG30ST
Only null left
Specifications
Capacity (Metric) 30 L
Color Black
Material LDPE, Polyethylene
Dimensions (L x W) 50 x 70cm
Length (Metric) 50 cm
Height (Metric) 70 cm
Weight (Metric) 8 kg
Product Type Waste Bag