Learn More
R&D Systems™ Reconstitution Buffer 7 (Deionized Water)
Product Code. 30000917
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000917 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 30000917 Supplier R&D Systems™ Supplier No. RB07
  • ONLINE EXCLUSIVE PRICE
    56.00€
    52.50€ / 2mL
    Save 3.50€
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
This product is intended as a companion product for reconstitution of selected lyophilized antibody or protein to maximize recovery and activity.
Store the unopened product at 2 - 8°C. Do not use past expiration date.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 mL