missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Reconstitution Buffer 6 (BSA/Acetic Acid)

Product Code. 30001069
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001069 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001069 Supplier R&D Systems™ Supplier No. RB06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

0.1% Bovine Serum Albumin in 100 mM Acetic Acid

This product is intended as a companion product for reconstitution of selected lyophilized antibody or protein to maximize recovery and activity. Refer to the specific antibody or protein product data sheet for the recommended reconstitution concentrations.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.