Learn More
R&D Systems™ Reconstitution Buffer 2 (BSA/PBS)
Product Code. 30002147
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002147 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30002147 Supplier R&D Systems™ Supplier No. RB02
0.1% Bovine Serum Albumin in PBS
This product is intended as a companion product for reconstitution of selected lyophilized antibody or protein to maximize recovery and activity. Refer to the specific antibody or protein product data sheet for the recommended reconstitution concentrations.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 mL