missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Nicotinamidase Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Nicotinamidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7235-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_011478.3 | |
S. cerevisiae Nicotinamidase Protein | |
Research Use Only | |
PNC1 | |
Recombinant Protein | |
E. coli | |
MKTLIVVDMQ NDFISPLGSL TVPKGEELIN PISDLMQDAD RDWHRIVVTR DWHPSRHISF AKNHKDKEPY STYTYHSPRP GDDSTQEGIL WPVHCVKNTW GSQLVDQIMD QVVTKHIKIV DKGFLTDREY YSAFHDIWNF HKTDMNKYLE KHHTDEVYIV GVALEYCVKA TAISAAELGY KTTVLLDYTR PISDDPEVIN KVKEELKAHN INVVDK | |
Greater than 90% as determined by SDS-PAGE. |
852846 | |
10 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |