missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Glycerol 2-dehydrogenase (NADP(+)) Protein
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Latent membrane protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7055-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_014763.1 | |
S. cerevisiae Glycerol 2-dehydrogenase (NADP(+)) Protein | |
Research Use Only | |
GCY1 | |
Recombinant Protein | |
E. coli | |
MPATLHDSTK ILSLNTGAQI PQIGLGTWQS KENDAYKAVL TALKDGYRHI DTAAIYRNED QVGQAIKDSG VPREEIFVTT KLWCTQHHEP EVALDQSLKR LGLDYVDLYL MHWPARLDPA YIKNEDILSV PTKKDGSRAV DITNWNFIKT WELMQELPKT GKTKAVGVSN FSINNLKDLL ASQGNKLTPA ANQVEIHPLL PQDELINFCK SKGIVVEAYS PLGSTDAPLL KEPVILEIAK KNNVQPGHVV ISWHVQRGYV VLPKSVNPDR IKTNRKIFTL STEDFEAINN ISKEKGEKRV VHPNWSPFEV FK | |
Greater than 90% as determined by SDS-PAGE. |
854287 | |
500 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |