missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae Cystathionine beta-lyase Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Cystathionine beta-lyase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7238-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037820

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_011331.3
S. cerevisiae Cystathionine beta-lyase Protein
Research Use Only
STR3
Recombinant Protein
E. coli
MPIKRLDTVV VNTGSQNDQH SASVPPVYLS TTFKVDLNNE DAQNYDYSRS GNPTRSVLQH QIGKLYRVPQ ENVLAVSSGM TALDVILRGL VLLNGTDNHT PTIIAGDDLY GGTQRLLNFF KQQSHAVSVH VDTSDFEKFK TVFQSLDKVD CVLLESPTNP LCKVVDIPRI LRFVKCISPD TTVVVDNTMM SGLNCNPLQL NPGCDVVYES ATKYLNGHHD LMGGVIISKT PEIASKLYFV INSTGAGLSP MDSWLLVRGL KTLGVRLYQQ QRNAMILAHW LENSCGFKPT RTNKATKTRF VGLRSNPDFK LHKSFNNGPG AVLSFETGSF EHSKRLVSSK KLSIWAVTVS FGCVNSLLSM PCKMSHASID PELRKERDFP EDLVRLCCGI ENIVDLKKDL LAAMVDADII EVRENGKYLF NKLNKNLAVN TTIDDLHKPL SIYEEFYNQD LIRKDSELNI KSSKL
Greater than 90% as determined by SDS-PAGE.
852691
500 μg
Not determined.
Full Length
S. cerevisiae
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers