missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Alpha-galactosidase 1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Alpha-galactosidase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7286-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
S. cerevisiae Alpha-galactosidase 1 Protein | |
Research Use Only | |
MEL1 | |
Recombinant Protein | |
E. coli | |
VSPSYNGLGL TPQMGWDNWN TFACDVSEQL LLDTADRISD LGLKDMGYKY IILDDCWSSG RDSDGFLVAD EQKFPNGMGH VADHLHNNSF LFGMYSSAGE YTCAGYPGSL GREEEDAQFF ANNRVDYLKY DNCYNKGQFG TPEISYHRYK AMSDALNKTG RPIFYSLCNW GQDLTFYWGS GIANSWRMSG DVTAEFTRPD SRCPCDGDEY DCKYAGFHCS IMNILNKAAP MGQNAGVGGW NDLDNLEVGV GNLTDDEEKA HFSMWAMVKS PLIIGANVNN LKASSYSIYS QASVIAINQD SNGIPATRVW RYYVSDTDEY GQGEIQMWSG PLDNGDQVVA LLNGGSVSRP MNTTLEEIFF DSNLGSKKLT STWDIYDLWA NRVDNSTASA ILGRNKTATG ILYNATEQSY KDGLSKNDTR LFGQKIGSLS PNAILNTTVP AHGIAFYRLR PSS | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |