missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae A-agglutinin-binding subunit Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) A-agglutinin-binding subunit, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7176-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027570

  • 3405.00€ / 1mg
Estimated Shipment:
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_011483.3
S. cerevisiae A-agglutinin-binding subunit Protein
Research Use Only
AGA2
Recombinant Protein
E. coli
QELTTICEQI PSPTLESTPY SLSTTTILAN GKAMQGVFEY YKSVTFVSNC GSHPSTTSKG SPINTQYVF
Greater than 90% as determined by SDS-PAGE.
852851
1 mg
Not determined.
Full Length
S. cerevisiae
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers