missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus fabF Protein

A DNA sequence encoding the Staphylococcus aureus (strain COL) fabF, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7667-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020081

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

WP_000081240.1
S. aureus fabF Protein
Research Use Only
fabF
Recombinant Protein
E. coli
MSQNKRVVIT GMGALSPIGN DVKTTWENAL KGVNGIDKIT RIDTEPYSVH LAGELKNFNI EDHIDKKEAR RMDRFTQYAI VAAREAVKDA QLDINENTAD RIGVWIGSGI GGMETFEIAH KQLMDKGPRR VSPFFVPMLI PDMATGQVSI DLGAKGPNGA TVTACATGTN SIGEAFKIVQ RGDADAMITG GTEAPITHMA IAGFSASRAL STNDDIETAC RPFQEGRDGF VMGEGAGILV IESLESAQAR GANIYAEIVG YGTTGDAYHI TAPAPEGEGG SRAMQAAMDD AGIEPKDVQY LNAHGTSTPV GDLNEVKAIK NTFGEAAKHL KVSSTKSMTG HLLGATGGIE AIFSALSIKD SKVAPTIHAV TPDPECDLDI VPNEAQDLDI TYAMSNSLGF GGHNAVLVFK KFEA
Greater than 90% as determined by SDS-PAGE.
31213734
10 μg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers