missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Enterotoxin type C-2 Protein
A DNA sequence encoding the Staphylococcus aureus Enterotoxin type C-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7197-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_001043552.1 | |
50 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
S. aureus Enterotoxin type C-2 Protein | |
Research Use Only | |
entC2 | |
Recombinant Protein | |
E. coli | |
ESQPDPTPDE LHKSSEFTGT MGNMKYLYDD HYVSATKVMS VDKFLAHDLI YNISDKKLKN YDKVKTELLN EDLAKKYKDE VVDVYGSNYY VNCYFSSKDN VGKVTGGKTC MYGGITKHEG NHFDNGNLQN VLIRVYENKR NTISFEVQTD KKSVTAQELD IKARNFLINK KNLYEFNSSP YETGYIKFIE NNGNTFWYDM MPAPGDKFDQ SKYLMMYNDN KTVDSKSVKI EVHLTTKNG | |
Greater than 90% as determined by SDS-PAGE. |