missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Adapter protein MecA Protein

A DNA sequence encoding the Staphylococcus aureus (strain MW2) Adapter protein MecA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7252-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017860

  • 985.00€ / 100µg
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

WP_001217728.1
S. aureus Adapter protein MecA Protein
Research Use Only
mecA
Recombinant Protein
E. coli
MRIERVDDTT VKLFITYSDI EARGFSREDL WTNRKRGEEF FWSMMDEINE EEDFVVEGPL WIQVHAFEKG VEVTISKSKN EDMMNMSDDD ATDQFDEQVQ ELLAQTLEGE DQLEELFEQR TKEKEAQGSK RQKSSARKNT RTIIVKFNDL EDVINYAYHS NPITTEFEDL LYMVDGTYYY AVHFDSHVDQ EVINDSYSQL LEFAYPTDRT EVYLNDYAKI IMSHNVTAQV RRYFPETTE
Greater than 90% as determined by SDS-PAGE.
31213749
100 μg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers