missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus 50S ribosomal protein L7 / L12 Protein
A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) 50S ribosomal protein L7/L12, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7259-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
WP_001273586.1 | |
S. aureus 50S ribosomal protein L7 / L12 Protein | |
Research Use Only | |
rplL | |
Recombinant Protein | |
E. coli | |
MANHEQIIEA IKEMSVLELN DLVKAIEEEF GVTAAAPVAV AGAAGGADAA AEKTEFDVEL TSAGSSKIKV VKAVKEATGL GLKDAKELVD GAPKVIKEAL PKEEAEKLKE QLEEVGATVE LK | |
Greater than 90% as determined by SDS-PAGE. |
31213346 | |
200 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |