missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rotavirus Intermediate capsid protein VP6 Protein

A DNA sequence encoding the Rotavirus A (strain Cow/France/RF/1975 G6-P6[1]-I2-R2-C2-M2-A3-N2-T6-E2-H3) (RV-A) Intermediate capsid protein VP6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7063-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007290

  • 1765.00€ / 200µg
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Rotavirus Intermediate capsid protein VP6 Protein
Research Use Only
VP6
Recombinant Protein
E. coli
MDVLYSLSKT LKDARDKIVE GTLYSNVSDL IQQFNQMIIT MNGNEFQTGG IGNLPIRNWN FDFGLLGTTL LNLDANYVET ARNTIDYFVD FVDNVCMDEM VRESQRNGIA PQSDSLIKLS GIKFKRINFD NSSEYIENWN LQNRRQRTGF TFHKPNIFPY SASFTLNRSQ PAHDNLMGTM WLNAGSEIQV AGFDYSCAIN APANTQQFEH IVQLRRVLTT ATITLLPDAE RFSFPRVITS ADGATTWYFN PVILRPNNVE IEFLLNGQII NTYQARFGTI IARNFDTIRL SFQLMRPPNM TPAVAALFPN AQPFEHHATV GLTLRIESAV CESVLADASE TMLANVTSVR QEYAIPVGPV FPPGMNWTDL ITNYSPSRED NLQRVFTVAS IRSMLVK
Greater than 90% as determined by SDS-PAGE.
200 μg
Not determined.
Full Length
Rotavirus
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers