missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Zinc transporter ZIP13 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Zinc transporter ZIP13, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7638-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001034285.1 | |
Zinc transporter ZIP13 Protein | |
Research Use Only | |
Slc39a13 | |
Recombinant Protein | |
E. coli | |
WAYTCNISPG VEGQSLQRQQ QLGLWVIAGF LTFLALEKMF LNCKEEDPSQ APSKDPTAAA LNGGHCLAQP AAEPGLRAVV RNLKVSGYLN LLANTIDNFT HGLA | |
Greater than 90% as determined by SDS-PAGE. |
295928 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His-GST | |
Tris-based buffer, 50% glycerol |