missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Tryptophan 2,3-dioxygenase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Tryptophan 2,3-dioxygenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6771-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_071798.1 | |
Tryptophan 2,3-dioxygenase Protein | |
Research Use Only | |
Tdo2 | |
Recombinant Protein | |
E. coli | |
MSGCPFSGNS VGYTLKNLSM EDNEEDGAQT GVNRASKGGL IYGDYLQLEK ILNAQELQSE IKGNKIHDEH LFIITHQAYE LWFKQILWEL DSVREIFQNG HVRDERNMLK VMTRMHRVVV IFKLLVQQFS VLETMTALDF NDFREYLSPA SGFQSLQFRL LENKIGVLQS LRVPYNRKHY RDNFEGDYNE LLLKSEQEQT LLQLVEAWLE RTPGLEPHGF NFWGKFEKNI LKGLEEEFLK IQAKKDSEEK EEQMAEFRKQ KEVLLCLFDE KRHDYLLSKG ERRLSYRALQ GALMIYFYRE EPRFQVPFQL LTSLMDIDTL MTKWRYNHVC MVHRMLGSKA GTGGSSGYYY LRSTVSDRYK VFVDLFNLSS YLVPRHWIPK MNPIIHKFLY TAEYSDSSYF SSDESD | |
Greater than 90% as determined by SDS-PAGE. |
64206 | |
500 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |