missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Suppressor of cytokine signaling 3 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Suppressor of cytokine signaling 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6713-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035310

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_446017.1
Suppressor of cytokine signaling 3 Protein
Research Use Only
Socs3
Recombinant Protein
E. coli
MVTHSKFPAA GMSRPLDTSL RLKTFSSKSE YQLVVNAVRK LQESGFYWSA VTGGEANLLL SAEPAGTFLI RDSSDQRHFF TLSVETQSGT KNLRIQCEGG SFSLQSDPRS TQPVPRFDCV LKLVHHYMPP PGAPSFSLPP TEPSFEVQEQ PPAQALPGGT PKRAYYIYSG GEKIPLVLSR PLSSNVATLQ HLCRKTVNGH LDSYEKVTQL PGPIREFLDQ YDAPL
Greater than 90% as determined by SDS-PAGE.
89829
500 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers