missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Sortilin Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Sortilin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6716-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16025340

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_113955.1
Sortilin Protein
Research Use Only
Sort1
Recombinant Protein
E. coli
CEENDYTTWL AHSTDPGDYK DGCILGYKEQ FLRLRKSSVC QNGRDYVVAK QPSICPCSLE DFLCDFGYFR PENASECVEQ PELKGHELEF CLYGKEEHLT TNGYRKIPGD RCQGGMNPAR EVKDLKKKCT SNFLNPKKQN SKSSS
Greater than 90% as determined by SDS-PAGE.
83576
200 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers