missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Nqo2 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Nqo2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7703-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16070291

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001004214.1
Nqo2 Protein
Research Use Only
Nqo2
Recombinant Protein
E. coli
MAGKKVLLVY AHQEPKSFNG SMKQVAVEEL SKQGCTVTVS DLYTMNFEPR ATRNDVTGAL SNPEVFKYGI EAYEAYKKKA LTSDILEEQR KVQEADLVIF QFPLYWFSVP AILKGWMDRV LCQGFAFDVP GFYDSGFLKD KLALLSFTTG GTAEMYTKAG VNGDFRYFLW PLQHGTLHFC GFKVLAPQIS FGPEVSSEEQ RKVMLASWVQ RLKSIWKEEP IHCTPSWYFQ G
Greater than 90% as determined by SDS-PAGE.
291084
200 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers