missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat NADPH oxidase 4 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) NADPH oxidase 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9309-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_445976.1 | |
NADPH oxidase 4 Protein | |
Research Use Only | |
Nox4 | |
Recombinant Protein | |
Yeast | |
GGLLKYQTNL DTHPPGCISL NRTPSQNMSI ADYVSEHFHG SLPGGFSKLE DHYQKTLVKI CLEEPKFQAH FPQTWIWISG PLCLYCAERL YRCIRSNKPV TIISVINHPS DVMELRMIKE NFKARPGQYI ILHCPSVSAL ENHPFTLTMC PTETKATFGV HFKVVGDWTE RFRDLLLPPS SQDSEILPFI QSRNYPKLYI DGPFGSPFEE SLNYE | |
Greater than 90% as determined by SDS-PAGE. |
85431 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |