missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IGF1 / IGF-I Protein

A DNA sequence encoding the Rattus norvegicus (Rat) IGF1 / IGF-I, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8580-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035401

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001075946.2
IGF1 / IGF-I Protein
Research Use Only
IGF1
Recombinant Protein
E. coli
GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSA
Greater than 90% as determined by SDS-PAGE.
24482
500 μg
Not determined.
Full Length
Rat
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers