missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Glutathione S-transferase P Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Glutathione S-transferase P, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6127-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051010

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_036709.1
Glutathione S-transferase P Protein
Research Use Only
Gstp1
Recombinant Protein
E. coli
PPYTIVYFPV RGRCEATRML LADQGQSWKE EVVTIDVWLQ GSLKSTCLYG QLPKFEDGDL TLYQSNAILR HLGRSLGLYG KDQKEAALVD MVNDGVEDLR CKYGTLIYTN YENGKDDYVK ALPGHLKPFE TLLSQNQGGK AFIVGNQISF ADYNLLDLLL VHQVLAPGCL DNFPLLSAYV ARLSARPKIK AFLSSPDHLN RPINGNGKQ
Greater than 90% as determined by SDS-PAGE.
24426
500 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers